Learn More
Biolegend Biotin anti-mouse LAP (TGF-β1)
Supplier:  Biolegend 141411
Encompass_Preferred
Biotin anti-mouse LAP (TGF-β1) [TW7-16B4]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: ELISA Detection; Size: 50 ug
Catalog No. NC1736968