Learn More
Cytek Biosciences Inc Antibodies Biotin Anti-Mouse CD16 / CD32 (2.4G2) ..... Cat. No. 30-0161-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 300161U025
Encompass_Preferred
Antibodies Biotin Anti-Mouse CD16 / CD32 (2.4G2) ..... Cat. No. 30-0161-U025 ..... Size 25 µgThe 2.4G2 antibody is specific for a common epitope found in the extracellular regions of mouse Fc-receptors Fc-gamma II (CD32) and Fc-gamma III (CD16). These receptors are expressed by B cells, monocytes, macrophages, NK cells, dendritic cells, and neutrophils.
Catalog No. 50-210-3153