Learn More
Cytek Biosciences Inc Antibodies Biotin Anti-Mouse CD45 (30-F11) ..... Cat. No. 30-0451-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 300451U025
Encompass_Preferred
Antibodies Biotin Anti-Mouse CD45 (30-F11) ..... Cat. No. 30-0451-U025 ..... Size 25 µgThe 30-F11 antibody reacts with mouse CD45, which is one of the most abundant hematopoietic markers and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). CD45 is a protein tyrosine phosphatase existing in several isoforms, each being generated and expressed in cell-specific patterns. With its broad cell distribution, CD45 is critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA, CD45RB, CD45RO and others.The 30-F11 antibody is widely used as a leukocyte marker for B cells, T cell subsets and NK cell subsets.
Catalog No. 50-210-3164