Learn More
Biolegend Biotin anti-mouse IL-9
Supplier:  Biolegend 531704
Encompass_Preferred
Biotin anti-mouse IL-9 [Poly5317]; Isotype: Goat Polyclonal Ig; Reactivity: Mouse; Apps: ELISA Detection; Size: 50 ug
Catalog No. 50-208-5978