Learn More
Biolegend Biotin anti-mouse Ly-6C [HK1.4]; Isotype: Rat IgG2c, ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Supplier:  Biolegend 128003
Encompass_Preferred
Biotin anti-mouse Ly-6C [HK1.4]; Isotype: Rat IgG2c, ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-164-431