Learn More
Biolegend Biotin anti-mouse CCL28 (MEC)
Supplier:  Biolegend 524203
Encompass_Preferred
Biotin anti-mouse CCL28 (MEC) [Poly5242]; Isotype: Goat Polyclonal Ig; Reactivity: Mouse; Apps: ELISA Detection; Size: 50 ug
Catalog No. 50-208-5943