Learn More
Biolegend Biotin anti-mouse CD104
Supplier:  Biolegend 123603
Encompass_Preferred
Biotin anti-mouse CD104 [346-11A]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-604-963