Learn More
Biolegend Biotin anti-mouse CD117 (c-kit)
Supplier:  Biolegend 105803
Encompass_Preferred
Biotin anti-mouse CD117 (c-kit) [2B8]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-163-086