Learn More
Biolegend Biotin anti-mouse CD11c
Supplier:  Biolegend 117304
Encompass_Preferred
Biotin anti-mouse CD11c [N418]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, IF; Size: 500 ug
Catalog No. NC9594905