Learn More
Biolegend Biotin anti-mouse CD122 (IL-2Rβ)
Supplier:  Biolegend 105904
Encompass_Preferred
Biotin anti-mouse CD122 (IL-2Rβ) [5H4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-163-107