Learn More
Biolegend Biotin anti-mouse CD138 (Syndecan-1)
Supplier:  Biolegend 142512
Encompass_Preferred
Biotin anti-mouse CD138 (Syndecan-1) [281-2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-207-1394