Learn More
Biolegend Biotin anti-mouse CD152
Supplier:  Biolegend 106303
Encompass_Preferred
Biotin anti-mouse CD152 [UC10-4B9]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-163-122