Learn More
Biolegend Biotin anti-mouse CD16/32
Supplier:  Biolegend 101303
Encompass_Preferred
Biotin anti-mouse CD16/32 [93]; Isotype: Rat IgG2a, λ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-162-540