Learn More
Biolegend Biotin anti-mouse CD183 (CXCR3)
Supplier:  Biolegend 126503
Encompass_Preferred
Biotin anti-mouse CD183 (CXCR3) [CXCR3-173]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-164-300