Learn More
Biolegend Biotin anti-mouse CD19
Supplier:  Biolegend 101504
Encompass_Preferred
Biotin anti-mouse CD19 [MB19-1]; Isotype: Mouse IgA, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-162-567