Learn More
Biolegend Biotin anti-mouse CD195 (CCR5)
Supplier:  Biolegend 107003
Encompass_Preferred
Biotin anti-mouse CD195 (CCR5) [HM-CCR5]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-163-146