Learn More
Biolegend Biotin anti-mouse CD197 (CCR7)
Supplier:  Biolegend 120103
Encompass_Preferred
Biotin anti-mouse CD197 (CCR7) [4B12]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-163-825