Learn More
Biolegend Biotin anti-mouse CD23
Supplier:  Biolegend 101604
Encompass_Preferred
Biotin anti-mouse CD23 [B3B4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-162-572