Learn More
Biolegend Biotin anti-mouse CD24
Supplier:  Biolegend 101803
Encompass_Preferred
Biotin anti-mouse CD24 [M1/69]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-162-586