Learn More
Biolegend Biotin anti-mouse CD28
Supplier:  Biolegend 102104
Encompass_Preferred
Biotin anti-mouse CD28 [37.51]; Isotype: Syrian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-162-639