Learn More
Biolegend Biotin anti-mouse CD326 (Ep-CAM)
Supplier:  Biolegend 118203
Encompass_Preferred
Biotin anti-mouse CD326 (Ep-CAM) [G8.8]; Isotype: Rat IgG2a, κ; Reactivity: mouse; Apps: FC; Size: 50 ug
Catalog No. 50-163-755