Learn More
Biolegend Biotin anti-mouse CD34
Supplier:  Biolegend 119304
Encompass_Preferred
Biotin anti-mouse CD34 [MEC14.7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 500 ug
Catalog No. 50-163-791