Learn More
Biolegend Biotin anti-mouse CD4
Supplier:  Biolegend 100508
Encompass_Preferred
Biotin anti-mouse CD4 [RM4-5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-604-780