Learn More
Biolegend Biotin anti-mouse CD45.2
Supplier:  Biolegend 109803
Encompass_Preferred
Biotin anti-mouse CD45.2 [104]; Isotype: Mouse (SJL) IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-163-367