Learn More
Biolegend Biotin anti-mouse CD45
Supplier:  Biolegend 103104
Encompass_Preferred
Biotin anti-mouse CD45 [30-F11]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-162-776