Learn More
Biolegend Biotin anti-mouse CD49d
Supplier:  Biolegend 103703
Encompass_Preferred
Biotin anti-mouse CD49d [9C10 (MFR4.B)]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-162-874