Learn More
Biolegend Biotin anti-mouse CD5
Supplier:  Biolegend 100604
Encompass_Preferred
Biotin anti-mouse CD5 [53-7.3]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-162-462