Learn More
Biolegend Biotin anti-mouse CD62L
Supplier:  Biolegend 104404
Encompass_Preferred
Biotin anti-mouse CD62L [MEL-14]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-206-9814