Learn More
Biolegend Biotin anti-mouse CD73
Supplier:  Biolegend 127203
Encompass_Preferred
Biotin anti-mouse CD73 [TY/11.8]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-164-348