Learn More
Biolegend Biotin anti-mouse CD80
Supplier:  Biolegend 104703
Encompass_Preferred
Biotin anti-mouse CD80 [16-10A1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Cross-Reactivity: Dog (Canine); Apps: FC; Size: 50 ug
Catalog No. 50-162-965