Learn More
Biolegend Biotin anti-mouse CD88 (C5aR)
Supplier:  Biolegend 135811
Encompass_Preferred
Biotin anti-mouse CD88 (C5aR) [20/70]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-1147