Learn More
Biolegend Biotin anti-mouse CD8a
Supplier:  Biolegend 100703
Encompass_Preferred
Biotin anti-mouse CD8a [53-6.7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC; Size: 50 ug
Catalog No. 50-162-475