Learn More
Biolegend Biotin anti-mouse FcεRIα
Supplier:  Biolegend 134303
Encompass_Preferred
Biotin anti-mouse FcεRIα [MAR-1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-164-728