Learn More
Biolegend Biotin anti-mouse GM-CSF
Supplier:  Biolegend 505502
Encompass_Preferred
Biotin anti-mouse GM-CSF [MP1-31G6]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ELISA Detection, ELISPOT Detection; Size: 500 ug
Catalog No. 50-169-905