Learn More
Biolegend Biotin anti-mouse H-2Kb
Supplier:  Biolegend 116504
Encompass_Preferred
Biotin anti-mouse H-2Kb [AF6-88.5]; Isotype: Mouse (BALB/c) IgG2a, κ; Reactivity: Mouse b haplotype; Apps: FC; Size: 500 ug
Catalog No. 50-163-665