Learn More
Biolegend Biotin anti-mouse/human NGF
Supplier:  Biolegend 509802
Encompass_Preferred
Biotin anti-mouse/human NGF [JKhNGF-2]; Isotype: Mouse IgG1; Reactivity: Mouse, Human, 2.5S, 7S, and β-NGF forms Cross-Reactivity: Cattle (Bovine, Cow), Viper; Apps: ELISA Detection; Size: 500 ug
Catalog No. 50-170-069