Learn More
Biolegend Biotin anti-mouse I-A/I-E
Supplier:  Biolegend 107603
Encompass_Preferred
Biotin anti-mouse I-A/I-E [M5/114.15.2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-163-176