Learn More
Biolegend Biotin anti-mouse IgMa
Supplier:  Biolegend 408603
Encompass_Preferred
Biotin anti-mouse IgMa [MA-69]; Isotype: Mouse (C57BL/6) IgG1, κ; Reactivity: Mouse; Apps: ELISA Detection, FC, IHC; Size: 50 ug
Catalog No. 50-169-235