Learn More
Biolegend Biotin anti-mouse IL-12/IL-23 p40
Supplier:  Biolegend 505301
Encompass_Preferred
Biotin anti-mouse IL-12/IL-23 p40 [C17.8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, IL-12/IL-23 p40 subunit (monomer, homodimer and heterodimer IL-12 p35/p40 or IL-23 p19/p40); Apps: ELISA Detection, ELISPOT Detection, ICFC; Size: 50 ug
Catalog No. 50-169-891