Learn More
Biolegend Biotin anti-mouse IL-1α
Supplier:  Biolegend 512503
Encompass_Preferred
Biotin anti-mouse IL-1α [BL1a-89]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ELISA Detection, ELISPOT Detection, WB, IP; Size: 50 ug
Catalog No. 50-170-137