Learn More
Biolegend Biotin anti-mouse IL-6
Supplier:  Biolegend 504601
Encompass_Preferred
Biotin anti-mouse IL-6 [MP5-32C11]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ELISA Detection, ELISPOT Detection; Size: 50 ug
Catalog No. 50-169-856