Learn More
Biolegend Biotin anti-mouse MAdCAM-1
Supplier:  Biolegend 120705
Encompass_Preferred
Biotin anti-mouse MAdCAM-1 [MECA-367]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-163-872