Learn More
Biolegend Biotin anti-mouse NK-1.1
Supplier:  Biolegend 108703
Encompass_Preferred
Biotin anti-mouse NK-1.1 [PK136]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-163-280