Learn More
Biolegend Biotin anti-mouse/rat TNF-α
Supplier:  Biolegend 516003
Encompass_Preferred
Biotin anti-mouse/rat TNF-α [Poly5160]; Isotype: Goat Polyclonal IgG; Reactivity: Mouse, Rat; Apps: ELISA Detection; Size: 50 ug
Catalog No. 50-208-5891