Learn More
Biolegend Biotin anti-mouse TCR γ/δ
Supplier:  Biolegend 118103
Encompass_Preferred
Biotin anti-mouse TCR γ/δ [GL3]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-163-744