Learn More
Biolegend Biotin anti-Neurofilament H (NF-H), Phosphorylated
Supplier:  Biolegend 801608
Encompass_Preferred
Biotin anti-Neurofilament H (NF-H), Phosphorylated [SMI 31]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse, Rat; Apps: IHC-P, WB; Size: 100 ug
Catalog No. 50-208-9852