Learn More
Biolegend Biotin anti-human/mouse/rat PCNA
Supplier:  Biolegend 307904
Encompass_Preferred
Biotin anti-human/mouse/rat PCNA [PC10]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse, Rat, Cross-Reactivity: Other species; Apps: ICFC; Size: 100 ug
Catalog No. 50-166-448