Learn More
Biolegend Biotin anti-rat IFN-γ
Supplier:  Biolegend 518803
Encompass_Preferred
Biotin anti-rat IFN-γ [Poly5188]; Isotype: Goat Polyclonal Ig; Reactivity: Rat; Apps: ELISA Detection, WB; Size: 50 ug
Catalog No. 50-605-540