Learn More
Biolegend Biotin anti-rat IgG2c [MRG2c-67]; Isotype: Mouse IgG; Reactivity: Rat; Apps: ELISA, FC; Size: 50 ug
Supplier:  Biolegend 407603
Encompass_Preferred
Biotin anti-rat IgG2c [MRG2c-67]; Isotype: Mouse IgG; Reactivity: Rat; Apps: ELISA, FC; Size: 50 ug
Catalog No. 50-169-211