Learn More
Biolegend Biotin anti-Synapsin I/II/III [A17080A]; Isotype: Mouse IgG2b, ; Reactivity: Human, Rat; Apps: WB, IHC-P; Size: 100 ug
Supplier:  Biolegend 853706
Encompass_Preferred
Biotin anti-Synapsin I/II/III [A17080A]; Isotype: Mouse IgG2b, ; Reactivity: Human, Rat; Apps: WB, IHC-P; Size: 100 ug
Catalog No. 50-207-4716